Learn More
Aviva Systems Biology Corp Pnpla6 Antibody - C-terminal region (ARP46767_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46767_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against Pnpla6. It was validated on Western Blot. The immunogen is a synthetic peptide corresponding to a region of Mouse. Alternate names: AI661849, MSws, NteApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-272