Learn More
Abcam pNPP-Na he x ahydrate, Alkaline phosphatase substrate, 5G
Catalog No. 502704847
Encompass_Preferred
MW 371.15 g/mol, Purity >98%. Chromogenic substrate for acid and alkaline phosphatases. The reaction produces a soluble end product that is yellow in color. The reaction may be stopped with 3 N NaOH and read at 405 nm. Can be used to perform kinetic and endpoint determinations.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4847 Supplier Abcam Supplier No. AB1462035G
MW 371.15 g/mol, Purity >98%. Chromogenic substrate for acid and alkaline phosphatases. The reaction produces a soluble end product that is yellow in color. The reaction may be stopped with 3 N NaOH and read at 405 nm. Can be used to perform kinetic and endpoint determinations.
The product is subject to the following: Abcam Restricted Use Statement