Learn More
Cayman Chemical pNPP Tablets 25ea
SDP
Catalog No. 502978353
Encompass_Preferred
A substrate for all alkaline phosphatase-based ELISA kits.
Catalog No. 50-297-8353 Supplier Cayman Chemical Supplier No. 40009025 EA
A substrate for all alkaline phosphatase-based ELISA kits.