Learn More
Cayman Chemical pNPP Tablets 5ea
SDP
Catalog No. 502942900
Encompass_Preferred
A substrate for all alkaline phosphatase-based ELISA kits.
Catalog No. 50-294-2900 Supplier Cayman Chemical Supplier No. 4000905 EA
A substrate for all alkaline phosphatase-based ELISA kits.