Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PNR-7-02 | 1633660-76-0 | 98.0% | 445.92 | C24H16ClN3O2S | 10 MG
SDP

Catalog No. 5000227782
Encompass_Preferred

PNR-7-02 is a small-molecule Pol η (DNA polymerase eta) inhibitor provided for preclinical biochemical and cell-based research. It is supplied as a solid research compound and is not for human or veterinary use.

  • Purity 98.0%.
  • Molecular weight 445.92 g/mol.
  • Chemical formula C24H16ClN3O2S.
  • CAS number 1633660-76-0.
  • Soluble in dimethyl sulfoxide (DMSO).
  • Pack size 10 mg.
  • Intended for research use only.

Catalog No. 50-002-27782 Supplier Medchemexpress LLC Supplier No. HY11766410MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PNR-7-02 is a small-molecule Pol η (DNA polymerase eta) inhibitor provided for preclinical biochemical and cell-based research. It is supplied as a solid research compound and is not for human or veterinary use.

  • Purity 98.0%.
  • Molecular weight 445.92 g/mol.
  • Chemical formula C24H16ClN3O2S.
  • CAS number 1633660-76-0.
  • Soluble in dimethyl sulfoxide (DMSO).
  • Pack size 10 mg.
  • Intended for research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.