Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PNR-7-02 | 1633660-76-0 | 98.0% | 445.92 | C24H16ClN3O2S | 5 MG
SDP

Catalog No. 5000226244
Encompass_Preferred

A small-molecule inhibitor of DNA polymerase eta (Pol η) supplied for research use. Provided as a solid in milligram quantities for in vitro and preclinical studies, it is suitable for mechanism-of-action experiments and combination cytotoxicity assays.

  • High purity: 98.0%.
  • Molecular weight 445.92.
  • Chemical formula C24H16ClN3O2S.
  • Provided in a 5 MG vial suitable for laboratory assays.
  • Intended for in vitro pharmacology and mechanism-of-action studies.

Catalog No. 50-002-26244 Supplier Medchemexpress LLC Supplier No. HY1176645MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

A small-molecule inhibitor of DNA polymerase eta (Pol η) supplied for research use. Provided as a solid in milligram quantities for in vitro and preclinical studies, it is suitable for mechanism-of-action experiments and combination cytotoxicity assays.

  • High purity: 98.0%.
  • Molecular weight 445.92.
  • Chemical formula C24H16ClN3O2S.
  • Provided in a 5 MG vial suitable for laboratory assays.
  • Intended for in vitro pharmacology and mechanism-of-action studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.