Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PNR-7-02 | 1633660-76-0 | 98.0% | 445.92 | C24H16ClN3O2S | 50 MG
SDP

Catalog No. 5000226243
Encompass_Preferred

PNR-7-02 is a small-molecule research inhibitor of DNA polymerase eta (Pol η). It is an indole thio-barbituric acid derivative reported to inhibit Pol η (IC50 ≈ 8 μM), with activity against hRev1 and DNA polymerase λ, and reported synergy with cisplatin in certain cancer cell lines.

  • Purity 98.0%.
  • Molecular weight 445.92.
  • Chemical formula C24H16ClN3O2S.
  • Appearance solid.
  • Recommended solvent DMSO; solubility note: 1 mg/mL, ultrasonic and warming, heat to 60°C.
  • Storage powder: -20°C for 3 years; in solvent: -80°C for 6 months.

Catalog No. 50-002-26243 Supplier Medchemexpress LLC Supplier No. HY11766450MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

PNR-7-02 is a small-molecule research inhibitor of DNA polymerase eta (Pol η). It is an indole thio-barbituric acid derivative reported to inhibit Pol η (IC50 ≈ 8 μM), with activity against hRev1 and DNA polymerase λ, and reported synergy with cisplatin in certain cancer cell lines.

  • Purity 98.0%.
  • Molecular weight 445.92.
  • Chemical formula C24H16ClN3O2S.
  • Appearance solid.
  • Recommended solvent DMSO; solubility note: 1 mg/mL, ultrasonic and warming, heat to 60°C.
  • Storage powder: -20°C for 3 years; in solvent: -80°C for 6 months.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.