Learn More
Medchemexpress LLC PNR-7-02 | 1633660-76-0 | 98.0% | 445.92 g·mol⁻¹ | C24H16ClN3O2S | 25 MG
SDP
Catalog No. 5000227783
Encompass_Preferred
PNR-7-02 is a small-molecule inhibitor of DNA polymerase eta (Pol η) used in research on translesion synthesis and polymerase-mediated drug resistance. It is an indole thio-barbituric acid derivative reported to inhibit Pol η (IC50 ≈ 8 μM) and to act synergistically with cisplatin in cancer cell lines.
  • Inhibits Pol η with reported IC50 ≈ 8 μM
  • Also shows activity against hRev1 and hpol λ
  • High purity (98.0%) suitable for biochemical assays
  • Available as a 25 MG package, other sizes available
  • Indole thio-barbituric acid scaffold, useful as a small-molecule probe
  • For research use only; not for human or clinical use
Catalog No. 50-002-27783 Supplier Medchemexpress LLC Supplier No. HY11766425MG
PNR-7-02 is a small-molecule inhibitor of DNA polymerase eta (Pol η) used in research on translesion synthesis and polymerase-mediated drug resistance. It is an indole thio-barbituric acid derivative reported to inhibit Pol η (IC50 ≈ 8 μM) and to act synergistically with cisplatin in cancer cell lines.
  • Inhibits Pol η with reported IC50 ≈ 8 μM
  • Also shows activity against hRev1 and hpol λ
  • High purity (98.0%) suitable for biochemical assays
  • Available as a 25 MG package, other sizes available
  • Indole thio-barbituric acid scaffold, useful as a small-molecule probe
  • For research use only; not for human or clinical use