Learn More
Aviva Systems Biology Corp PNRC1 Antibody - middle region (ARP52260_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52260_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against PNRC1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human PNRC1. Alternate names: B4-2, PNAS-145, PROL2, PRR2, RP11-63L7.5ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-285-677