Learn More
Cayman Chemical PntoIc AcId 10mg
SDP
Supplier:  Cayman Chemical 3668510 MG
Encompass_Preferred
An intermediate in the biosynthesis of D-pantothenic acid and, subsequently, CoA
Catalog No. 50-291-3249