Learn More
Cayman Chemical Pantoic Acid, Purity: ≥90%, CAS: 1112-33-0, 50 mg
SDP
Catalog No. 502910071
Encompass_Preferred
An intermediate in the biosynthesis of D-pantothenic acid and, subsequently, CoA
Catalog No. 50-291-0071 Supplier Cayman Chemical Supplier No. 3668550 MG
An intermediate in the biosynthesis of D-pantothenic acid and, subsequently, CoA