Learn More
Cayman Chemical Pntothenat KInase InHBTr 10mg
SDP
Catalog No. 502988382
Encompass_Preferred
A reversible inhibitor of pantothenate kinase (PanK; IC50s = 70, 92, and 25 nM for PanK1β, PanK2, and PanK3, respectively); inhibits CoA biosynthesis in C3A cells (IC50 = 0.9 µM); sensitizes cells to ferroptosis inducers
Catalog No. 50-298-8382 Supplier Cayman Chemical Supplier No. 3100210 MG
Add to Cart
Add to Cart
A reversible inhibitor of pantothenate kinase (PanK; IC50s = 70, 92, and 25 nM for PanK1β, PanK2, and PanK3, respectively); inhibits CoA biosynthesis in C3A cells (IC50 = 0.9 µM); sensitizes cells to ferroptosis inducers