Learn More
Cayman Chemical Pantothenate Kinase Inhibitor, Purity: ≥95%, CAS: 902614-04-4, 1 mg
SDP
Catalog No. 502989639
Encompass_Preferred
A reversible inhibitor of pantothenate kinase (PanK; IC50s = 70, 92, and 25 nM for PanK1β, PanK2, and PanK3, respectively); inhibits CoA biosynthesis in C3A cells (IC50 = 0.9 µM); sensitizes cells to ferroptosis inducers
Catalog No. 50-298-9639 Supplier Cayman Chemical Supplier No. 310021 MG
A reversible inhibitor of pantothenate kinase (PanK; IC50s = 70, 92, and 25 nM for PanK1β, PanK2, and PanK3, respectively); inhibits CoA biosynthesis in C3A cells (IC50 = 0.9 µM); sensitizes cells to ferroptosis inducers