Learn More
Cayman Chemical Pantothenate-AMC, Purity: ≥95%, CAS: 1224691-12-6, 1 mg
SDP
Catalog No. 502895941
Encompass_Preferred
A fluorogenic substrate for vanin-1; AMC is released upon enzymatic cleavage by vanin-1 and its fluorescence can be used to quantify vanin-1 activity; ex/em = 340-360/440-460 nm, respectively
Catalog No. 50-289-5941 Supplier Cayman Chemical Supplier No. 90044631 MG
A fluorogenic substrate for vanin-1; AMC is released upon enzymatic cleavage by vanin-1 and its fluorescence can be used to quantify vanin-1 activity; ex/em = 340-360/440-460 nm, respectively