Learn More
Medchemexpress LLC PNU-100440 | 168828-82-8 | MFCD06658241 | 99.7% | 296.29 g/mol | C14H17FN2O4 | 25 G
SDP
Catalog No. 5000303874
Encompass_Preferred
PNU-100440 is an endogenous metabolite of the antibiotic linezolid supplied as a research-grade reference compound for in vitro and preclinical studies. It is a white to off-white solid with molecular formula C14H17FN2O4 and molecular weight 296.29 g/mol, offered in solid and DMSO solution formats for analytical and biological research.
  • Endogenous metabolite of linezolid, useful as a reference standard.
  • High purity of 99.7% suitable for analytical applications.
  • Available as solid and DMSO solution to support assay workflows.
  • White to off-white solid with defined physical properties.
  • Characterized by standard identifiers including CAS and MFCD numbers.
Catalog No. 50-003-03874 Supplier Medchemexpress LLC Supplier No. 25GHYW019700
PNU-100440 is an endogenous metabolite of the antibiotic linezolid supplied as a research-grade reference compound for in vitro and preclinical studies. It is a white to off-white solid with molecular formula C14H17FN2O4 and molecular weight 296.29 g/mol, offered in solid and DMSO solution formats for analytical and biological research.
  • Endogenous metabolite of linezolid, useful as a reference standard.
  • High purity of 99.7% suitable for analytical applications.
  • Available as solid and DMSO solution to support assay workflows.
  • White to off-white solid with defined physical properties.
  • Characterized by standard identifiers including CAS and MFCD numbers.