Learn More
Enzo Life Sciences PNU-120596 (5mg). CAS: 501925-31-1
SDP
Supplier:  Enzo Life Sciences ALX550526M005
Encompass_Preferred
Type II positive allosteric modulator (PAM) of α7 neuronal nicotinic acetylcholine receptor (nAChR) (EC50=216nM). Shows no detectable effect on α4β2, α3β4 and α9α10 receptors. Alternative name: NSC 216666, N-(5-Chloro-2,4-dimethoxyphenyl)-N'-(5-methyl-3-isoxazolyl)urea. Purity: ≥98% (HPLC). Appearance: White to off-white solid. Solubility: Soluble in DMSO (50mg/ml); slightly soluble in methanol (1mg/ml) or acetonitrile (0.5mg/ml). Long Term Storage: -20°C.
Catalog No. 50-200-9561