Learn More
Abcam PNU 120596, Positive allosteric modulator for alpha7 nicotinic acetylcholine receptors, 10MG
Catalog No. 502707487
Encompass_Preferred
MW 311.72 Da, Purity >99%. Competitive, type II positive allosteric modulator for α7nAChR (EC₅₀ = 216 nM). Increases potency of nicotinic agonists. No effect on α4β2, α3β4 and α9α10 nAChRs. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7487 Supplier Abcam Supplier No. AB12055910MG
MW 311.72 Da, Purity >99%. Competitive, type II positive allosteric modulator for α7nAChR (EC₅₀ = 216 nM). Increases potency of nicotinic agonists. No effect on α4β2, α3β4 and α9α10 nAChRs. Active in vivo.
The product is subject to the following: Abcam Restricted Use Statement