Learn More
TARGETMOL CHEMICALS INC PNU-177864 HYDROCHLORIDE 25MG
SDP
Catalog No. 502577423
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PNU-177864 hydrochloride is a potent selective and orally active antagonist of dopamine D3 receptor. PNU-177864 hydrochloride is structurally consistent with a cationic amphiphilic drug (CAD). PNU-177864 hydrochloride induces phospholipidosis in vivo. PNU-177864 hydrochloride exhibits antischizophrenic activity. purity: 99%
Catalog No. 50-257-7423 Supplier TARGETMOL CHEMICALS INC Supplier No. T1251025MG
Also available in 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PNU-177864 hydrochloride is a potent selective and orally active antagonist of dopamine D3 receptor. PNU-177864 hydrochloride is structurally consistent with a cationic amphiphilic drug (CAD). PNU-177864 hydrochloride induces phospholipidosis in vivo. PNU-177864 hydrochloride exhibits antischizophrenic activity. purity: 99%