Learn More
Medchemexpress LLC PNU-22394 hydrochloride | 15923-42-9 | 98.8% | 236.74 | C13H17ClN2 | 10 MG
SDP
Catalog No. 5000245794
Encompass_Preferred
PNU-22394 hydrochloride is a research-grade small molecule agonist of serotonin (5-HT) receptors, reported as a 5-HT2C agonist and partial 5-HT2A/2B agonist. Supplied as the hydrochloride salt and provided at high purity for pharmacology and receptor biology studies in vitro and in vivo.
  • Potent 5-HT2C agonist with reported Ki ≈ 6.1 nM and 5-HT2A activity ≈ 10 nM.
  • Hydrochloride salt, suitable for dissolution in common assay solvents.
  • Molecular formula C13H17ClN2 and molecular weight 236.74 g/mol.
  • High purity (>98.7%), appropriate for research applications.
  • Intended for pharmacological research and receptor characterization studies.
Catalog No. 50-002-45794 Supplier Medchemexpress LLC Supplier No. HY10314510MG
PNU-22394 hydrochloride is a research-grade small molecule agonist of serotonin (5-HT) receptors, reported as a 5-HT2C agonist and partial 5-HT2A/2B agonist. Supplied as the hydrochloride salt and provided at high purity for pharmacology and receptor biology studies in vitro and in vivo.
  • Potent 5-HT2C agonist with reported Ki ≈ 6.1 nM and 5-HT2A activity ≈ 10 nM.
  • Hydrochloride salt, suitable for dissolution in common assay solvents.
  • Molecular formula C13H17ClN2 and molecular weight 236.74 g/mol.
  • High purity (>98.7%), appropriate for research applications.
  • Intended for pharmacological research and receptor characterization studies.