Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PNU-22394 hydrochloride | 15923-42-9 | 98.8% | 236.74 g/mol | C13H17ClN2 | 5 MG
SDP

Catalog No. 5000247209
Encompass_Preferred

PNU-22394 hydrochloride is a research-grade small-molecule agonist of serotonin (5-HT) receptors, showing selectivity for 5-HT2C and activity at 5-HT2A. Supplied as a hydrochloride salt, it is intended for in vitro pharmacology and receptor signaling studies.

  • Selective agonist at 5-HT2C with Ki = 6.1 nM.
  • Also active at 5-HT2A (Ki = 10 nM).
  • Assay-dependent EC50: 5-HT2C 7.7 nM (Fluo-4/Ca2+) and 0.12 μM (IP-One); 5-HT2A 23 nM (Fluo-4/Ca2+) and 0.21 μM (IP-One).
  • Molecular formula C13H17ClN2; molecular weight 236.74 g/mol.
  • High chemical purity (98.8%).
  • Available in small milligram quantities for laboratory research.

Catalog No. 50-002-47209 Supplier Medchemexpress LLC Supplier No. HY1031455MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PNU-22394 hydrochloride is a research-grade small-molecule agonist of serotonin (5-HT) receptors, showing selectivity for 5-HT2C and activity at 5-HT2A. Supplied as a hydrochloride salt, it is intended for in vitro pharmacology and receptor signaling studies.

  • Selective agonist at 5-HT2C with Ki = 6.1 nM.
  • Also active at 5-HT2A (Ki = 10 nM).
  • Assay-dependent EC50: 5-HT2C 7.7 nM (Fluo-4/Ca2+) and 0.12 μM (IP-One); 5-HT2A 23 nM (Fluo-4/Ca2+) and 0.21 μM (IP-One).
  • Molecular formula C13H17ClN2; molecular weight 236.74 g/mol.
  • High chemical purity (98.8%).
  • Available in small milligram quantities for laboratory research.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.