Learn More
Enzo Life Sciences PNU-282987 (10mg). CAS: 123464-89-1
SDP
Supplier:  Enzo Life Sciences BMLC1200010
Encompass_Preferred
A highly selective α7 nicotinic acetylcholine receptor agonist and prototypical novel therapeutic for schizophrenia. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (20 mg/ml), water (6 mg/ml) or ethanol (6 mg/ml). Long Term Storage: Ambient.
Catalog No. 50-201-0345