Learn More
Abcam PNU 282987, alpha7 nAChR agonist, 50MG
Catalog No. 502707488
Encompass_Preferred
MW 301.2 Da, Purity >99%. Selective α7nAChR agonist. Negligible activity at α1β1γδ and α3β4 receptors. High affinity for rat α7nACh receptors (Ki = 26 nM). Functional antagonist at 5-HT₃ receptors (IC₅₀ = 4541 nM).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7488 Supplier Abcam Supplier No. AB12055850MG
MW 301.2 Da, Purity >99%. Selective α7nAChR agonist. Negligible activity at α1β1γδ and α3β4 receptors. High affinity for rat α7nACh receptors (Ki = 26 nM). Functional antagonist at 5-HT₃ receptors (IC₅₀ = 4541 nM).
The product is subject to the following: Abcam Restricted Use Statement