Learn More
Selleck Chemical LLC PNU 282987 HCl
SDP
Catalog No. 5000002838
Encompass_Preferred
PNU 282987 is a potent agonist of 7-containing neuronal nicotinic acetylcholine receptors with Ki value of 26 nM for the rat receptor
Catalog No. 50-000-02838 Supplier Selleck Chemical LLC Supplier No. S59305MG
Add to Cart
Add to Cart
PNU 282987 is a potent agonist of 7-containing neuronal nicotinic acetylcholine receptors with Ki value of 26 nM for the rat receptor