Learn More
Enzo Life Sciences PNU-74654 (5 mg). CAS: 113906-27-7
SDP
Supplier:  Enzo Life Sciences BMLWN1010005
Encompass_Preferred
Anti-tumor agent that disrupts the canonical Wnt/β-catenin pathway via blockage of the protein-protein interaction between β-catenin and TCF. Alternative name: 2-Phenoxybenzoic acid [(5-methyl-2-furanyl)methylene]hydrazide. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO (25 mg/ml) and in 100% ethanol (8 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-2359