Learn More
Medchemexpress LLC HY-101130 5mg Medchemexpress, PNU-74654 CAS:113906-27-7 Purity:>98%
SDP
Catalog No. 501932048
Encompass_Preferred
Medchemexpress, HY-101130 5mg PNU-74654 CAS:113906-27-7 PNU-74654 is an inhibitor of Wnt/β-catenin pathway with an IC 50 of 129.8 μM in NCI-H295 cell. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2048 Supplier Medchemexpress LLC Supplier No. HY1011305MG
Medchemexpress, HY-101130 5mg PNU-74654 CAS:113906-27-7 PNU-74654 is an inhibitor of Wnt/β-catenin pathway with an IC 50 of 129.8 μM in NCI-H295 cell. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.