Learn More
Abcam PNU 74654, Wnt signaling pathway blocker, 25MG
Catalog No. 502705567
Encompass_Preferred
MW 320.3 Da, Purity >98%. Wnt signaling pathway blocker. Inhibits Wnt/β-catenin pathway. Blocks the protein-protein interaction between β-catenin and TCF. Antitumor effects.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5567 Supplier Abcam Supplier No. AB14461325MG
MW 320.3 Da, Purity >98%. Wnt signaling pathway blocker. Inhibits Wnt/β-catenin pathway. Blocks the protein-protein interaction between β-catenin and TCF. Antitumor effects.
The product is subject to the following: Abcam Restricted Use Statement