Learn More
TARGETMOL CHEMICALS INC PNU112455A HYDROCHLORIDE 10MG
SDP
Catalog No. 502576255
Encompass_Preferred
Also available in 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PNU112455A hydrochloride is an ATP site competetive inhibitor of CDK2 and CDK5binds to the ATP site of CDK2 and CDK5 with Kms of 3.6 and 3.2 uM respectively. purity: 100%
Catalog No. 50-257-6255 Supplier TARGETMOL CHEMICALS INC Supplier No. T823010MG
Also available in 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PNU112455A hydrochloride is an ATP site competetive inhibitor of CDK2 and CDK5binds to the ATP site of CDK2 and CDK5 with Kms of 3.6 and 3.2 uM respectively. purity: 100%