Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC PNU112455A hydrochloride | 21886-12-4 | 99.6% | 301.75 | 1 ML
SDP

Catalog No. 5000435853
Encompass_Preferred

PNU112455A hydrochloride is an ATP-competitive inhibitor of CDK2 and CDK5. It binds to the ATP site of CDK2 and CDK5 with Km values of 3.6 μM and 3.2 μM, respectively. This product is intended for research use only and is not sold to patients.

  • ATP-competitive CDK2 and CDK5 inhibitor.
  • Binds to the ATP site of CDK2 and CDK5.
  • Km for CDK2: 3.6 μM.
  • Km for CDK5: 3.2 μM.
  • Purity: 99.56%.
  • Appearance: Solid, Light yellow to yellow.
  • Shipping: Room temperature in continental US.
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month.

Catalog No. 50-004-35853 Supplier Medchemexpress LLC Supplier No. HY11246810MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PNU112455A hydrochloride is an ATP-competitive inhibitor of CDK2 and CDK5. It binds to the ATP site of CDK2 and CDK5 with Km values of 3.6 μM and 3.2 μM, respectively. This product is intended for research use only and is not sold to patients.

  • ATP-competitive CDK2 and CDK5 inhibitor.
  • Binds to the ATP site of CDK2 and CDK5.
  • Km for CDK2: 3.6 μM.
  • Km for CDK5: 3.2 μM.
  • Purity: 99.56%.
  • Appearance: Solid, Light yellow to yellow.
  • Shipping: Room temperature in continental US.
  • Storage: 4°C, sealed storage, away from moisture. In solvent: -80°C for 6 months; -20°C for 1 month.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.