Learn More
Cayman Chemical PnumocndIn B0 10mg
SDP
Catalog No. 502987490
Encompass_Preferred
An antifungal lipopeptide that inhibits the synthesis of β-(1,3)-D-glucan, a component of fungal cell walls (IC50s = 70 and 67 ng/ml for inhibiting glucan synthase in C. albicans and A. fumigatus, respectively)
Catalog No. 50-298-7490 Supplier Cayman Chemical Supplier No. 2152710 MG
An antifungal lipopeptide that inhibits the synthesis of β-(1,3)-D-glucan, a component of fungal cell walls (IC50s = 70 and 67 ng/ml for inhibiting glucan synthase in C. albicans and A. fumigatus, respectively)