Learn More
TARGETMOL CHEMICALS INC PNZ5 5MG
SDP
Catalog No. 502581446
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PNZ5 is a potent and isoxazole-based pan-BET inhibitor with high selectivity and potency similar to the well-established (+)-JQ1 with a KD of 5.43 nM for BRD4(1). purity: 99%
Catalog No. 50-258-1446 Supplier TARGETMOL CHEMICALS INC Supplier No. T125135MG
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. PNZ5 is a potent and isoxazole-based pan-BET inhibitor with high selectivity and potency similar to the well-established (+)-JQ1 with a KD of 5.43 nM for BRD4(1). purity: 99%