Learn More
SAI Infusion Technologies Rodent Oral Gavage (Feeding) Needle - 20G 38mm with Stainless Steel Bite-Guard
SDP
Catalog No. NC1525822
Encompass
20G 38mm Flexible and Disposable Feeding Needles with a Stainless Steel Bite-Guard for maximum durability. SAI's smooth needle tip and flexible PTFE tubing prevent damage to the esophagus during dosing, while the unique Bite-Guard prevents any biting damage to the needle. These needles are sterile and packed 5/pk, 20 packs/box. Product is for laboratory animal research only.
Catalog No. NC1525822 Supplier SAI Infusion Technologies Supplier No. FN2038M
 Restricted Item
Add to Cart
Add to Cart
20G 38mm Flexible and Disposable Feeding Needles with a Stainless Steel Bite-Guard for maximum durability. SAI's smooth needle tip and flexible PTFE tubing prevent damage to the esophagus during dosing, while the unique Bite-Guard prevents any biting damage to the needle. These needles are sterile and packed 5/pk, 20 packs/box. Product is for laboratory animal research only.