Learn More
Abcam PO-PRO3, 200UL
Supplier:  Abcam AB275918200UL
Encompass_Preferred
MW 641.42 g/mol. Cell-impermeant, high affinity nucleic acid stain. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-268-2740