Learn More
Nova Biomedical Corporation PO2 Electrode, for Use in Bioprofile Flex
Catalog No. NC9067466
Encompass
PO2 Electrode for use in Bioprofile Flex. Measures 3.0-800.0 mmHg range and 0.1 mmHg resolution via Clarke Electrode.
Catalog No. NC9067466 Supplier Nova Biomedical Corporation Supplier No. 42456
PO2 Electrode for use in Bioprofile Flex. Measures 3.0-800.0 mmHg range and 0.1 mmHg resolution via Clarke Electrode.