Learn More
Medchemexpress LLC POBN 10mg | 66893-81-0 | 10 MG
SDP
Catalog No. 5000228308
Encompass_Preferred
POBN (alpha-(4-pyridyl-1-oxide)-N-tert-butylnitrone) is a cell-permeable, hydrophilic spin-trap reagent used to detect free radical adducts in biochemical research. It is supplied at high purity for laboratory research applications.
  • Cell-permeable spin trap for detecting free radical adducts.
  • Hydrophilic, suitable for use in aqueous systems.
  • High purity (99.94%).
  • Provided for research use only.
  • Packaged as 10 mg aliquots for bench-scale experiments.
Catalog No. 50-002-28308 Supplier Medchemexpress LLC Supplier No. HY11471310MG
POBN (alpha-(4-pyridyl-1-oxide)-N-tert-butylnitrone) is a cell-permeable, hydrophilic spin-trap reagent used to detect free radical adducts in biochemical research. It is supplied at high purity for laboratory research applications.
  • Cell-permeable spin trap for detecting free radical adducts.
  • Hydrophilic, suitable for use in aqueous systems.
  • High purity (99.94%).
  • Provided for research use only.
  • Packaged as 10 mg aliquots for bench-scale experiments.