Learn More
Medchemexpress LLC POBN 50mg | 66893-81-0 | 50 MG
SDP
Catalog No. 5000229145
Encompass_Preferred
POBN (alpha-(4-pyridyl-1-oxide)-N-tert-butylnitrone) is a cell-permeable, hydrophilic nitrone spin trap used in research to detect and stabilize free radical adducts for analytical and biological studies. It is supplied as a reagent-grade solid for laboratory use and intended for research use only.
  • Cell-permeable nitrone spin trap for detecting free radicals.
  • Hydrophilic and soluble in polar solvents.
  • Stabilizes radical adducts for electron paramagnetic resonance and analytical detection.
  • Suitable for biochemical and chemical radical-trapping studies.
  • Provided as a reagent-grade solid for laboratory research.
Catalog No. 50-002-29145 Supplier Medchemexpress LLC Supplier No. HY11471350MG
POBN (alpha-(4-pyridyl-1-oxide)-N-tert-butylnitrone) is a cell-permeable, hydrophilic nitrone spin trap used in research to detect and stabilize free radical adducts for analytical and biological studies. It is supplied as a reagent-grade solid for laboratory use and intended for research use only.
  • Cell-permeable nitrone spin trap for detecting free radicals.
  • Hydrophilic and soluble in polar solvents.
  • Stabilizes radical adducts for electron paramagnetic resonance and analytical detection.
  • Suitable for biochemical and chemical radical-trapping studies.
  • Provided as a reagent-grade solid for laboratory research.