Learn More
Roche Diagnostics POC Chemstrip Urine Test Strips
Catalog No. BC417145 Shop All Roche Diagnostics Products
Change view
Click to view available options
Detectable Analytes:
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Urobilinogen
Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein
Blood, Glucose, Leukocyte, Nitrite, Protein
Glucose, Protein
Leukocyte, Nitrite
Type:
Chemstrip 10 MD
Chemstrip 10 SG
Chemstrip 2 GP
Chemstrip 2 LN
Chemstrip 5 OB
Chemstrip 7
Chemstrip 9
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes Type
BC417145 Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen Chemstrip 10 SG
22-045-620 Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein Chemstrip 7
22-045-621 Blood, Glucose, Leukocyte, Nitrite, Protein Chemstrip 5 OB
BC417109 Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Urobilinogen Chemstrip 9
BC200743 Glucose, Protein Chemstrip 2 GP
22-208-840 Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen Chemstrip 10 MD
BC417152 Leukocyte, Nitrite Chemstrip 2 LN
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. BC417145 Supplier Roche Diagnostics Supplier No. 11895362160
Add to Cart
Add to Cart
This item is not returnable. View return policy
Accuracy, performance and testing simplicity
  • Avoid potentially costly consequences of missed diagnoses — vitamin C interference is virtually eliminated on the glucose pad1, 2
  • Avoid false negatives — both acetoacetic acid and acetone are detected on the ketone reagent pad3
  • Simplify and streamline reading — all reagent pads are read at one time, between one and two minutes after dipping3
  • Promote safe and hygienic testing practice — fingers stay clear of specimen
  • Generate confidence in test result accuracy — Chemstrip construction allows uniform liquid penetration and prevents runover onto adjacent pads
TRUSTED_SUSTAINABILITY
Specifications
Clia Complexity Waived
Detectable Analytes Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
Type Chemstrip 10 SG
For Use With (Application) Urinalysis
Quantity 100 Pack