Learn More
Perkin Elmer US LLC POC Injector with PPC Add-On Kit, 240 V
Catalog No. 502113738
Encompass_Preferred
Part of the Programmed-Temperature On-Column Injector System. The AutoSystem XL or Clarus GC must be PPC (Programmable Pneumatic Control) ready. If not, a PPC upgrade kit (N6120146) is required. Installation by PerkinElmer Service is required, but not included.
Catalog No. 50-211-3738 Supplier Perkin Elmer US LLC Supplier No. N6120143
Add to Cart
Add to Cart
Request a Quote
Part of the Programmed-Temperature On-Column Injector System. The AutoSystem XL or Clarus GC must be PPC (Programmable Pneumatic Control) ready. If not, a PPC upgrade kit (N6120146) is required. Installation by PerkinElmer Service is required, but not included.