Learn More
Perkin Elmer US LLC POC Packed Injector Assembly
Catalog No. 502113592
Encompass_Preferred
The Programmed-Temperature On-Column (POC) Inlet is designed for use with fused-silica capillary columns. The sample is injected onto the column while the inlet is cool. After the injection, the inlet begins to heat. This delay in heating avoids the flash vaporization associated with a normal injection. This explosive vaporization can cause thermal breakdown and/or discrimination of certain analytes, which can be avoided by using the POC. The POC Injector is best used to achieve recovery of compounds of greater than C60 such as polywaxes. The POC utilizes flow control, producing the best recovery out to C100 or greater.
Catalog No. 50-211-3592 Supplier Perkin Elmer US LLC Supplier No. N6100048
The Programmed-Temperature On-Column (POC) Inlet is designed for use with fused-silica capillary columns. The sample is injected onto the column while the inlet is cool. After the injection, the inlet begins to heat. This delay in heating avoids the flash vaporization associated with a normal injection. This explosive vaporization can cause thermal breakdown and/or discrimination of certain analytes, which can be avoided by using the POC. The POC Injector is best used to achieve recovery of compounds of greater than C60 such as polywaxes. The POC utilizes flow control, producing the best recovery out to C100 or greater.