Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kova Plastics by Alltrista POC Urinalysis System Pack

Catalog No. 22749802
Change view
Click to view available options
No. per Pack:
100 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
22-749-802 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 22-749-802 Supplier Kova Plastics by Alltrista Supplier No. 88100E

missing translation for 'validMainframeMsg'

Contains all components to standardize every step of the microscopic urinalysis process.

  • Convenient packaging for lower volume users
  • 100 Econo Tubes, Caps and Petters
  • 10 FastRead 102 Slides

Specifications

Sterility Non-sterile
For Use With (Application) Microscopic analysis of urine
Includes 100 Econo-Tubes, 100 Caps, 100 Petters, 10 FastRead 102 Slides
No. per Pack 100 Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.