Learn More
Medchemexpress LLC Pociredir hydrochloride | 2490676-19-0 | 99.9% | 439.87 | 1 MG
SDP
Catalog No. 5000349251
Encompass_Preferred
Pociredir hydrochloride (FTX-6058 hydrochloride) is a potent and orally active inhibitor of Embryonic Ectoderm Development (EED). It can induce HbF protein expression in cell and murine models and is used for research of select hemoglobinopathies, including sickle cell disease and β-thalassemia.
  • Potent and orally active inhibitor of EED.
  • Induces HbF protein expression in cell and murine models.
  • Used for research of select hemoglobinopathies, including sickle cell disease and β-thalassemia.
  • Inhibits PRC2 via binding to EED.
Catalog No. 50-003-49251 Supplier Medchemexpress LLC Supplier No. HY139400A1MG
Pociredir hydrochloride (FTX-6058 hydrochloride) is a potent and orally active inhibitor of Embryonic Ectoderm Development (EED). It can induce HbF protein expression in cell and murine models and is used for research of select hemoglobinopathies, including sickle cell disease and β-thalassemia.
  • Potent and orally active inhibitor of EED.
  • Induces HbF protein expression in cell and murine models.
  • Used for research of select hemoglobinopathies, including sickle cell disease and β-thalassemia.
  • Inhibits PRC2 via binding to EED.