Learn More
Medchemexpress LLC Pociredir hydrochloride | 2490676-19-0 | 99.9% | 439.87 | 10 MM/1 ML
SDP
Catalog No. 5000349264
Encompass_Preferred
Pociredir hydrochloride, also known as FTX-6058, is a potent and orally active inhibitor of Embryonic Ectoderm Development (EED). It is utilized in research for select hemoglobinopathies like sickle cell disease and β-thalassemia by inducing HbF protein expression in cell and murine models.
  • Potent and orally active inhibitor of EED
  • Induces HbF protein expression in cell and murine models
  • Used in research for select hemoglobinopathies, including sickle cell disease and β-thalassemia
  • Inhibits PRC2 via binding to EED
Catalog No. 50-003-49264 Supplier Medchemexpress LLC Supplier No. HY139400A10MM/1ML
Pociredir hydrochloride, also known as FTX-6058, is a potent and orally active inhibitor of Embryonic Ectoderm Development (EED). It is utilized in research for select hemoglobinopathies like sickle cell disease and β-thalassemia by inducing HbF protein expression in cell and murine models.
  • Potent and orally active inhibitor of EED
  • Induces HbF protein expression in cell and murine models
  • Used in research for select hemoglobinopathies, including sickle cell disease and β-thalassemia
  • Inhibits PRC2 via binding to EED