Learn More
Medchemexpress LLC Pociredir (hydrochloride) | 2490676-19-0 | 99.9% | 439.87 | 25 MG
SDP
Catalog No. 5000349914
Encompass_Preferred
Pociredir (FTX-6058) hydrochloride is a potent and orally active inhibitor of Embryonic Ectoderm Development (EED). It can induce HbF protein expression in cell and murine models and is used for the research of select hemoglobinopathies, including sickle cell disease and β-thalassemia.
  • Inhibits PRC2 via binding to EED
  • Induces robust HbF protein expression in cell and murine models
  • Used for research of select hemoglobinopathies
Catalog No. 50-003-49914 Supplier Medchemexpress LLC Supplier No. 25MGHY139400A
Pociredir (FTX-6058) hydrochloride is a potent and orally active inhibitor of Embryonic Ectoderm Development (EED). It can induce HbF protein expression in cell and murine models and is used for the research of select hemoglobinopathies, including sickle cell disease and β-thalassemia.
  • Inhibits PRC2 via binding to EED
  • Induces robust HbF protein expression in cell and murine models
  • Used for research of select hemoglobinopathies