Learn More
Medchemexpress LLC Pociredir hydrochloride | 2490676-19-0 | 99.9% | 439.87 | 5 MG
SDP
Catalog No. 5000364206
Encompass_Preferred
Pociredir hydrochloride (FTX-6058) is a potent and orally active inhibitor of Embryonic Ectoderm Development (EED). It can induce HbF protein expression in cell and murine models, making it suitable for research into select hemoglobinopathies, including sickle cell disease and β-thalassemia. It inhibits PRC2 by binding to EED, which leads to robust HbF protein expression.
  • Potent and orally active EED inhibitor
  • Induces HbF protein expression in cell and murine models
  • Useful for research on sickle cell disease and β-thalassemia
  • Inhibits PRC2 by binding to EED
Catalog No. 50-003-64206 Supplier Medchemexpress LLC Supplier No. 5MGHY139400A
Pociredir hydrochloride (FTX-6058) is a potent and orally active inhibitor of Embryonic Ectoderm Development (EED). It can induce HbF protein expression in cell and murine models, making it suitable for research into select hemoglobinopathies, including sickle cell disease and β-thalassemia. It inhibits PRC2 by binding to EED, which leads to robust HbF protein expression.
  • Potent and orally active EED inhibitor
  • Induces HbF protein expression in cell and murine models
  • Useful for research on sickle cell disease and β-thalassemia
  • Inhibits PRC2 by binding to EED