Learn More
Cole-Parmer Shimpo MT-100 Compact Contact Pocket/Handheld Tachometer
Supplier:  Cole-Parmer 9880719
Encompass
These rugged tachometers fit perfectly in the left or right hand, and are designed for use with a gloved hand. Units provide fourteen different units of measure including rpm, inch/minute, feet/minute to total length measurements like feet and meters, and more. Additional memory features include last, minimum, maximum, and average of results. Durable units are ideal for the most challenging industrial environments, and the compact design is small enough to fit into your shirt pocket. Model 98807-20 noncontact tachometer is easy to use and incorporates a laser that is accurate up to 6 ft (1.8 m) from the target, ensuring a safe distance from rotating parts.
Catalog No. 50-993-618