Learn More
Electron Microscopy Sciences Pocket Measuring Microscope 50x
SDP
Catalog No. 5027949
Encompass_Preferred
As small as a fountain pen, the pocket measuring microscope has a standard scale is incorporated in its lens barrel, giving an effecient brightness and excellent definition. It can be focused in the same way as a conventional microscope. Field of view: 2.00 mm. Min. scale division: 0.02. Measuring range: 1.60mm.
Size: 12.4 x 127 mm
Catalog No. 50-279-49 Supplier Electron Microscopy Sciences Supplier No. 68131
As small as a fountain pen, the pocket measuring microscope has a standard scale is incorporated in its lens barrel, giving an effecient brightness and excellent definition. It can be focused in the same way as a conventional microscope. Field of view: 2.00 mm. Min. scale division: 0.02. Measuring range: 1.60mm.
Size: 12.4 x 127 mm