Learn More
Electron Microscopy Sciences Pocket Mirror Stereo Viewer, 2x
SDP
Supplier:  Electron Microscopy Sciences 68100
Encompass_Preferred
A simple device, used for viewing a set of 3-dimensional aerial photos in 3-dimension. It comes with two support legs that are folded flat for storage and are unfolded for the viewing position. Photos are viewed through the two lenses simultaneously with both eyes. Lenses may be adjusted for suitable eye-to-eye width. The coated lenses are 32 mm in diameter. Magnifying power is two.
Catalog No. 50-291-28
Only null left
Edge