Learn More
Hach Company Pocket Pro Plus sample cup
Supplier:  Hach Company 9532900
Encompass
Pocket Pro Tester sample cup/cap for safe storage Can be used as sample cup / measurement beaker as well Does not fit to Pocket Pro Testers
Catalog No. 50-002-07220