Learn More
Cole-Parmer Atago PAL-39S Special-scale Refractometer, Hydrogen Peroxide
Catalog No. NC1573015 Shop All Cole-Parmer Products
Encompass_Preferred
These compact refractometers give you a direct reading scale of the concentration of some common liquids. Rugged IP65-rated design is ideal for indoor and field use. Simply apply two to three drops of sample onto the prism, press the start key, and the value is displayed in seconds. Digital display eliminates reading errors common with handheld refractometers. Cleanup is simple easily clean under running water.
Catalog No. NC1573015 Supplier Cole-Parmer Supplier No. 0294106
These compact refractometers give you a direct reading scale of the concentration of some common liquids. Rugged IP65-rated design is ideal for indoor and field use. Simply apply two to three drops of sample onto the prism, press the start key, and the value is displayed in seconds. Digital display eliminates reading errors common with handheld refractometers. Cleanup is simple easily clean under running water.