Learn More
Pharmacal Research Laboratories Pocket Swabs, ATP Rapid Hygiene Test, 5-Second Result Time, 100 Per Pack
SDP
Catalog No. NC1914250
Encompass
The PHARMACAL RESEARCH LABS Pocket Swabs are self-contained ATP rapid hygiene tests designed for accurate and swift surface cleanliness verification. Pre-filled with a biofilm breaking agent, these swabs ensure efficient sample collection and sensitivity in detecting microorganisms and residues. With immediate feedback in just 5 seconds, they enhance hygiene compliance in critical environments, such as food processing and healthcare. Their one-year shelf life and room temperature stability add to their convenience for diverse applications.
Catalog No. NC1914250 Supplier Pharmacal Research Laboratories Supplier No. 99NLPS100
The PHARMACAL RESEARCH LABS Pocket Swabs are self-contained ATP rapid hygiene tests designed for accurate and swift surface cleanliness verification. Pre-filled with a biofilm breaking agent, these swabs ensure efficient sample collection and sensitivity in detecting microorganisms and residues. With immediate feedback in just 5 seconds, they enhance hygiene compliance in critical environments, such as food processing and healthcare. Their one-year shelf life and room temperature stability add to their convenience for diverse applications.