Learn More
Cole-Parmer Pocket Thermometer; Range -58 to 302F (-50 to 150C)
Supplier:  Cole-Parmer 9009000
Encompass_Preferred
Cole-Parmer Pocket Thermometer; Range -58 to 302F (-50 to 150C), These thermometers are ideal for laboratory, field, and process measurements. Use to quickly verify temperature. Typical response time is 30 seconds for 99% of new reading. Thermometers have a 304 stainless steel penetration stem. All models include a protective case with pocket clip and a battery. Thermometer features a sealed caseideal for use with liquids. It has >F/>C switchable ranges, a MIN/MAX button, and a one-second display update. Use ON/OFF button to conserve battery life.
Catalog No. NC1662940